Order Entry
Belgium
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507

Biopsy processing/embedding cassettes, Micromesh™ M507

Pink
Catalog # 720-0972
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Catalog # 720-0972
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507

Pink

Price
518,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.

  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems

Documents

Specifications
  • Colour:
    Pink
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.