Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Biopsy processing/embedding cassettes, Micromesh™ M507

Turquoise
Catalog # 720-0970
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Catalog # 720-0970
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507

Turquoise

Price
518,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.

  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems

 

Specifications
  • Colour:
    Turquoise
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.