Order Entry
Belgium
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507

Biopsy processing/embedding cassettes, Micromesh™ M507

Orange
Catalog # 720-0969
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Catalog # 720-0969
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507

Orange

Price
518,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.

  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems

Documents

Specifications
  • Colour:
    Orange