Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Iron standard solution

Iron Standard Solution, 100 ml (NIST), 1000 mg/l as Fe, 100 mL
Catalog # HACH227142
Supplier:   Hach
Iron standard solution
Catalog # HACH227142
Supplier:  Hach
CAS Number:  
Iron standard solution
Iron standard solution

Iron Standard Solution, 100 ml (NIST), 1000 mg/l as Fe, 100 mL

CAS Number:  
Price
33,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

For accuracy checks in iron determinations.

  • Traceable to NIST
Specifications

Specifications

  • Concentration:
    1000 mg/l as Fe
  • Pk:
    100 mL
  • Description:
    Iron Standard Solution, 100 ml (NIST)

Specifications