Orders LinkContactUsLinkComponent
Lab Consumables
Lab Consumables
Aluminum loop holder with PVC handle, 175 mm, 100×1, in tube
Catalog # 612-2684
Supplier:   Copan
Lab Consumables
Catalog # 612-2684
Supplier:  Copan
Lab Consumables
Lab Consumables
Aluminum loop holder with PVC handle, 175 mm, 100×1, in tube
Price
39,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Specifications
  • Packed:
    100×1, in tube
  • Description:
    Aluminum loop holder with PVC handle, 175 mm
  • Seq:
    2