Orders LinkContactUsLinkComponent
VWR®, Slide Boxes
VWR®, Slide Boxes
Slide boxes, Red, PP, 25, 90×140×35 mm
Catalog # 631-0740
Supplier:   VWR Collection
VWR®, Slide Boxes
Catalog # 631-0740
Supplier:  VWR Collection
VWR®, Slide Boxes
VWR®, Slide Boxes
Slide boxes, Red, PP, 25, 90×140×35 mm
Price
9,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Slide boxes for holding standard microscopy slides.
  • Hinged lids held securely by a latch lock
  • Stacking feet maximise stability when placed on top of one another
  • Slide inventory sheet on the inside of the lid
W×D×H: 90×140×35 mm (for 25), 165×210×35 mm (for 100)
Specifications
  • No. of slides:
    25
  • Colour:
    Red
  • Description:
    Slide boxes
  • Material:
    PP
  • Seq:
    005
  • W×D×H:
    90×140×35 mm