VWR®, Slide Boxes
Slide boxes, Blue, PP, 25, 90×140×35 mm
Catalog # 631-0739
Supplier: VWR Collection
Jump to:
Product Details & Documents
Slide boxes for holding standard microscopy slides.
- Hinged lids held securely by a latch lock
- Stacking feet maximise stability when placed on top of one another
- Slide inventory sheet on the inside of the lid
W×D×H: 90×140×35 mm (for 25), 165×210×35 mm (for 100)
Documents
Specifications
- No. of slides:25
- Colour:Blue
- Description:Slide boxes
- Material:PP
- Seq:003
- W×D×H:90×140×35 mm