VWR®, Slide Boxes
Slide boxes, White, PP, 100, 165×210×35 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
Slide boxes for holding standard microscopy slides.
- Hinged lids held securely by a latch lock
- Stacking feet maximise stability when placed on top of one another
- Slide inventory sheet on the inside of the lid
W×D×H: 90×140×35 mm (for 25), 165×210×35 mm (for 100)
Documents
Specifications
- No. of slides:100
- Colour:White
- Description:Slide boxes
- Material:PP
- Seq:002
- W×D×H:165×210×35 mm