Orders LinkContactUsLinkComponent
VWR®, Microscope Slide Holders
VWR®, Microscope Slide Holders
Slide holder, White, PE, 178×299×19 mm, 78
Catalog # 631-0719
Supplier:   VWR Collection
VWR®, Microscope Slide Holders
Catalog # 631-0719
Supplier:  VWR Collection
VWR®, Microscope Slide Holders
VWR®, Microscope Slide Holders
Slide holder, White, PE, 178×299×19 mm, 78
Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sturdy microscope slide holder designed for drying and cleaning.
Colour: White
Documents
Specifications
  • No. of slides:
    78
  • Colour:
    White
  • Description:
    Slide holder
  • Material:
    PE
  • Seq:
    05
  • W×D×H:
    178×299×19 mm