BUFFER REDOX 468MV 30ML 1 * 6 items
Catalog # METN51319058
Supplier: Mettler - Toledo
Frequently Bought Together
VWR®, Pasteur Pipettes
Supplier: VWR Collection
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
BUFFER SOLN REDOX UH = 427MV 220MV 25°C 1 * 250 ml
Supplier: Mettler - Toledo