Order Entry
Belgium
Orders LinkContactUsLinkComponent
Precision probes, T

Precision probes, T

Stainless steel probe (IP 67) with PUR cable, −50…+350 °C, ±0,2 (−20…+70);
±1,0 °C, 4×110 mm
Catalog # 620-3317
Supplier:   TESTO
Precision probes, T
Catalog # 620-3317
Supplier:  TESTO
Precision probes, T
Precision probes, T

Stainless steel probe (IP 67) with PUR cable, −50…+350 °C, ±0,2 (−20…+70);
±1,0 °C, 4×110 mm

Price
95,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −50…+350 °C
  • Description:
    Stainless steel probe (IP 67) with PUR cable
  • Seq:
    03
  • Accuracy:
    ±0,2 (−20…+70);
    ±1,0 °C
  • Ø×L:
    4×110 mm