Orders LinkContactUsLinkComponent
pH probes
pH probes
Universal pH probe, plastic, for testo 206 pH3, with temperature sensor, 0 - 14, 0...80 °C, Gel, 12×120 mm, General purposes
Catalog # TEST0650.2064
Supplier:   TESTO
pH probes
Catalog # TEST0650.2064
Supplier:  TESTO
pH probes
pH probes
Universal pH probe, plastic, for testo 206 pH3, with temperature sensor, 0 - 14, 0...80 °C, Gel, 12×120 mm, General purposes
Price
224,00 €
Log in for availability
1 * 1 ST
Specifications
  • Ø×L:
    12×120 mm
  • For:
    General purposes
  • Description:
    Universal pH probe, plastic, for testo 206 pH3, with temperature sensor
  • Seq:
    04
  • Electrolyte:
    Gel
  • Temp.:
    0...80 °C
  • pH:
    0 - 14