pH probes
Universal pH probe, plastic, for testo 206 pH3, without temperature sensor, 0 - 14, - °C, Gel, 12×120 mm, General purposes
Catalog # TEST0650.2063
Supplier: TESTO
Jump to:
Specifications
- Ø×L:12×120 mm
- For:General purposes
- Description:Universal pH probe, plastic, for testo 206 pH3, without temperature sensor
- Seq:05
- Electrolyte:Gel
- Temp.:- °C
- pH:0 - 14