Orders LinkContactUsLinkComponent
Single-component explosives solutions
Single-component explosives solutions
Internal standard includes PGDN (propylene glycol dinitrate), with 1000 µg/ml in methanol, 1 ml/ampule, 1 Ampoul
Catalog # REST31821
Single-component explosives solutions
Catalog # REST31821
Supplier:  
CAS Number:  
Single-component explosives solutions
Single-component explosives solutions
Internal standard includes PGDN (propylene glycol dinitrate), with 1000 µg/ml in methanol, 1 ml/ampule, 1 Ampoul
CAS Number:  
Discontinued
Product Details & Documents
Single-component explosives solutions supports US department of defense base closures and remediation.
  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
Specifications
Specifications
  • Pk:
    1 Ampoul
  • Description:
    Internal standard includes PGDN (propylene glycol dinitrate), with 1000 µg/ml in methanol, 1 ml/ampule