Solid phase extraction cartridges, Resprep®, method specific
RDX method, for extraction of explosive compounds from water samples, PP, 6 ml, 500 mg
Catalog # REST26093
Supplier: Restek
Jump to:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifications
- Capacity:6 ml
- Description:RDX method, for extraction of explosive compounds from water samples
- Seq:04
- Bed weight:500 mg
- Column hardware:PP