Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Resprep®, C18 / C8
Resprep® SPE cartridge, high load, endcapped, Cartridge, C18, 1 ml, 100 mg
Catalog # REST26030
Supplier:   Restek
Solid phase extraction cartridges, Resprep®, C18 / C8
Catalog # REST26030
Supplier:  Restek
Solid phase extraction cartridges, Resprep®, C18 / C8
Solid phase extraction cartridges, Resprep®, C18 / C8
Resprep® SPE cartridge, high load, endcapped, Cartridge, C18, 1 ml, 100 mg
Price
414,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These cartridges are made of glass fiber disks embedded with C18 or C8 bonded silica. These hydrophobic (nonpolar) silica-based adsorbents are ideal to extract hydrophobic analytes from polar matrices, such as water (e.g., pesticides from water).
  • All cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted
  • Extract semivolatile organic compounds
  • Deep-pore design reduces clogging and allows faster flow rates
  • Lower cost than PTFE disks
 : Meet requirements for EPA Methods 525.1, 506, 550.1, and 549.1.
Specifications
  • Column Format:
    Cartridge
  • Capacity:
    1 ml
  • Description:
    Resprep® SPE cartridge, high load, endcapped
  • Seq:
    06
  • Bed weight:
    100 mg
  • Phase:
    C18