Order Entry
Belgium
Orders LinkContactUsLinkComponent
Shell vials

Shell vials

Shell vials, limited volume, 0,70 ml, 8×40 mm, PP, Clear, No
Catalog # REST24681
Supplier:   Restek
Shell vials
Catalog # REST24681
Supplier:  Restek
Shell vials
Shell vials

Shell vials, limited volume, 0,70 ml, 8×40 mm, PP, Clear, No

Price
234,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Specifications
  • Cap included:
    No
  • Ø×H:
    8×40 mm
  • Capacity:
    0,70 ml
  • Colour:
    Clear
  • Description:
    Shell vials, limited volume
  • Material:
    PP
  • Seq:
    08