Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Shell vials

Shell vials, 1,00 ml, 8×40 mm, Glass, Amber, No
  REST24664
 :   Restek
Shell vials
  REST24664
 :  Restek
Shell vials
Shell vials

Shell vials, 1,00 ml, 8×40 mm, Glass, Amber, No

 
40,20 €
   
1 * 200 ST
 
 
  • Cap included:
    No
  • Ø×H:
    8×40 mm
  • Capacity:
    1,00 ml
  • Colour:
    Amber
  • Description:
    Shell vials
  • Material:
    Glass
  • Seq:
    03