Flow controllers, Veriflo
8 – 15 ml/min, Stainless steel, Flow controller, 0,051 mm
REST24262
: Restek
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
- Ø:0,051 mm
- Maximum Flow Rate:8 – 15 ml/min
- Description:Flow controller
- Material:Stainless steel
- Seq:09



