Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Flow controllers, Veriflo

Flow controllers, Veriflo

2 – 4 ml/min, Acier inoxydable, Flow controller, 0,03 mm
Catalog # REST24260
Supplier:   Restek
Flow controllers, Veriflo
Catalog # REST24260
Supplier:  Restek
Flow controllers, Veriflo
Flow controllers, Veriflo

2 – 4 ml/min, Acier inoxydable, Flow controller, 0,03 mm

Price
1 280,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.

Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.

Specifications
  • Ø:
    0,03 mm
  • Débit maximal:
    2 – 4 ml/min
  • Désignation:
    Flow controller
  • Matière:
    Acier inoxydable
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.