Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Flow controllers, Veriflo

2 – 4 ml/min, Siltek® treated stainless steel, Flow controller, 0,03 mm
Catalog # REST24255
Supplier:   Restek
Flow controllers, Veriflo
Catalog # REST24255
Supplier:  Restek
Flow controllers, Veriflo
Flow controllers, Veriflo

2 – 4 ml/min, Siltek® treated stainless steel, Flow controller, 0,03 mm

Price
1 650,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.

Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.

Specifications
  • Ø:
    0,03 mm
  • Maximum Flow Rate:
    2 – 4 ml/min
  • Description:
    Flow controller
  • Material:
    Siltek® treated stainless steel
  • Seq:
    06