Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flow controllers, Veriflo

Flow controllers, Veriflo

15 – 30 ml/min, Siltek® treated stainless steel, Replacement orifice, 0,076 mm
Catalog # REST24236
Supplier:   Restek
Flow controllers, Veriflo
Catalog # REST24236
Supplier:  Restek
Flow controllers, Veriflo
Flow controllers, Veriflo

15 – 30 ml/min, Siltek® treated stainless steel, Replacement orifice, 0,076 mm

Price
340,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.

Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.

Specifications
  • Ø:
    0,076 mm
  • Maximum Flow Rate:
    15 – 30 ml/min
  • Description:
    Replacement orifice
  • Material:
    Siltek® treated stainless steel
  • Seq:
    110