Solid phase extraction cartridges, Resprep®, method specific
Organo tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bed, PP, 60 ml, -
Catalog # REST24049
Supplier: Restek
Jump to:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifications
- Capacity:60 ml
- Description:Organo tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bed
- Seq:06
- Bed weight:-
- Column hardware:PP