Protocol wall mounts for gas regulators
Protocol station, chrome-plated brass, CGA 350 (H2, P5) fitting
N° de catalogue REST21348
Fournisseur: Restek
Jump to:
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
- Ease of use
- Prevents gas regulator damage and improves safety
Either chrome-plated brass or 316 stainless steel option is complete with a 0,9m, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Specifications
- Description:Protocol station, chrome-plated brass, CGA 350 (H2, P5) fitting
- Seq:01