PIPES
Glass bottle for solids, 100 g
Catalog # MOLE37406078-100G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
- CAS Number:5625-37-6
- Formula:C₈H₁₈N₂O₆S₂
- MDL Number:MFCD00006159
- Molecular Weight:302.37 g/mol
- Merck Index:12,07633
- Boiling Pt:499 °C (1013 hPa)
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:227-057-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



