L(+)-Ascorbic acid
Plastic bottle for solids, 100 g
N° de catalogue MOLE35196085-100G
Marketsource
Fournisseur: Molekula
L(+)-Ascorbic acid
N° de catalogue MOLE35196085-100G
Fournisseur: Molekula
Marketsource
Numéro CAS: 50-81-7
Jump to:
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



