Colour comparison disk Ammonium suitable for Cat. No. 920006 VISOCOLOR HE. 1 * 1 items
Catalog # MANA920306
Supplier: MACHEREY-NAGEL
Colour comparison disk Ammonium suitable for Cat. No. 920006 VISOCOLOR HE. 1 * 1 items
Catalog # MANA920306
Supplier: MACHEREY-NAGEL
Frequently Bought Together
NEOMYCIN ELISA KIT 1 * 1 KIT
Supplier: ENZO LIFE SCIENCES
DCODE DEXTRAMER(10X)-GOLD HLA-B*07 1 * 25 Pce
Supplier: MARKET SOURCE PART PROCESS
SULFITES 10 MG/L IN H2O 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
EasySep™ RBC Depletion Reagents, 10 ml
Supplier: STEMCELL Technologies
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
VISOCOLOR® Colorimetric Test, Nitrite
Supplier: MACHEREY-NAGEL
CX-1 PH GRADIENT BUFFER B, PH 10.2, 1L 1 * 1 items
Supplier: Thermo Fisher Scientific
Silver Enhancer Kit, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
ACIDE 2,6-PYRIDINE CARBOXYLIQUE 0,02 M 1 * 10 L
Supplier: PROLABO CMP
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Supplier: Thermo Fisher Scientific
POTASSIUM CYANURE EN SOLUTION 1 * 100 mL
Supplier: PROLABO CMP
[EN]RAT IGG1 GL113 ISOTYPE CTRL LOWENDOT 1 * 5 mg
Supplier: MARKET SOURCE PART PROCESS
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
[EN]IGG4 ISOTYPE CTRL S228PF234AL235A LA 1 * 1 mg
Supplier: MARKET SOURCE PART PROCESS