Order Entry
Belgium
Orders LinkContactUsLinkComponent
Roller bottles, Nunc™ InVitro™

Roller bottles, Nunc™ InVitro™

Standard long (1XL), 1800 cm², 200-1000 ml
Catalog # 734-2395
   Low Manufacturing Impact
Roller bottles, Nunc™ InVitro™
Catalog # 734-2395
   Low Manufacturing Impact
Roller bottles, Nunc™ InVitro™
Roller bottles, Nunc™ InVitro™

Standard long (1XL), 1800 cm², 200-1000 ml

Price
381,00 €
Log in for availability
1 * 22 ST
Product Details & Documents

PETG, sterile.

  • Quick-action ergonomic closure reduces wrist stain and increases productivity
  • Wide range of sizes with surface areas from 1050 to 4200 cm²
  • Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
  • Easy-to-read graduations for medium fills
  • Lot number is printed on each bottle to maximise traceability

For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

Specifications
  • Growth area:
    1800 cm²
  • Recommended working volume:
    200-1000 ml
  • Description:
    Standard long (1XL)
  • Cat. No.:
    734-2395
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact