Order Entry
Belgium
Orders LinkContactUsLinkComponent
Micro-tissue grinders

Micro-tissue grinders

Type G 1 with pestle, 0,01 - 0,025 ml, ~4×30 mm
Catalog # ROTH0296.1
Marketsource
Supplier:   Roth Carl
Micro-tissue grinders
Catalog # ROTH0296.1
Supplier:  Roth Carl
Marketsource
Micro-tissue grinders

Type G 1 with pestle, 0,01 - 0,025 ml, ~4×30 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These micro-tissue grinders with pestles made of borosilicate glass are for pulverising and homogenising minute quantities.

Specifications
  • Int.Ø×H:
    ~4×30 mm
  • Capacity:
    0,01 - 0,025 ml
  • Description:
    Type G 1 with pestle
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.