Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

10 kg
Catalog # ROTH9356.2
Corrosive
Marketsource
Supplier:   Roth Carl
Sodium hydroxide
Catalog # ROTH9356.2
Supplier:  Roth Carl
Marketsource
CAS Number:  1310-73-2
Sodium hydroxide

10 kg

CAS Number:  1310-73-2
Price
142,50 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.