Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sodium nitrate

Glass bottle for solids, 2,5 kg
Catalog # ROTH8601.1
Oxidizing
Irritant
Marketsource
Supplier:   Roth Carl
Sodium nitrate
Catalog # ROTH8601.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7631-99-4
Sodium nitrate

Glass bottle for solids, 2,5 kg

CAS Number:  7631-99-4
Price
Inquire For Price
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:13,08720
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:308 °C
  • Density:2.26 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.