Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper

Copper

Glass bottle for solids, 1 kg
Catalog # ROTH8540.2
Marketsource
Supplier:   Roth Carl
Copper
Catalog # ROTH8540.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7440-50-8
Copper

Glass bottle for solids, 1 kg

CAS Number:  7440-50-8
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Merck Index:13,02545
  • Boiling Pt:2595 °C (1013 hPa)
  • Melting Pt:1083 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications