Potassium nitrate
Glass bottle for solids, 1 kg
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Nitrate of Potash
- CAS Number:7757-79-1
- Formula:KNO₃
- MDL Number:MFCD00011409
- Molecular Weight:101.1 g/mol
- Merck Index:13,07733
- Melting Pt:334 °C
- Density:2.109 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-818-8
- UN:1486
- ADR:5.1,III
- REACH:01-2119488224-35
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



