Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

30 L
Catalog # ROTHT172.5
Marketsource
Supplier:   Roth Carl
Water
Catalog # ROTHT172.5
Supplier:  Roth Carl
Marketsource
CAS Number:  7732-18-5
CAS Number:  7732-18-5
Price
169,00 €
Log in for availability
1 * 30 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pk:
    30 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.