Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

Glass bottle, 500 mL
Catalog # ROTHT143.2
Marketsource
Supplier:   Roth Carl
Water
Catalog # ROTHT143.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7732-18-5
Water

Glass bottle, 500 mL

CAS Number:  7732-18-5
Price
38,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.