Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methyl red

Methyl red

Glass bottle for solids, 25 g
Catalog # ROTHT120.2
Marketsource
Supplier:   Roth Carl
Methyl red
Catalog # ROTHT120.2
Supplier:  Roth Carl
Marketsource
CAS Number:  493-52-7
Methyl red

Glass bottle for solids, 25 g

CAS Number:  493-52-7
Price
35,90 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.