Lecithin
Glass bottle for solids, 1 kg
Catalog # ROTHAE81.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:8002-43-5
- MDL Number:MFCD00147406
- Merck Index:12,05452
- Density:1.0305 g/cm³ (20 °C)
- EINECS:232-307-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



