Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

5 L
Catalog # ROTHA511.3
Marketsource
   Safer Human and Environmental Health
Supplier:   Roth Carl
Water
Catalog # ROTHA511.3
Supplier:  Roth Carl
Marketsource
   Safer Human and Environmental Health
CAS Number:  7732-18-5
CAS Number:  7732-18-5
Price
81,50 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specifications

  • Pk:
    5 L
  • Cat. No.:
    ROTHA511.3
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.