Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium nitrate

Sodium nitrate

Glass bottle for solids, 500 g
Catalog # ROTHA136.1
Oxidizing
Irritant
Marketsource
Supplier:   Roth Carl
Sodium nitrate
Catalog # ROTHA136.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7631-99-4
Sodium nitrate

Glass bottle for solids, 500 g

CAS Number:  7631-99-4
Price
35,90 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:13,08720
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:308 °C
  • Density:2.26 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications