Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Ribose

D-(-)-Ribose

Glass bottle for solids, 100 g
Catalog # ROTH7168.2
Marketsource
Supplier:   Roth Carl
D-(-)-Ribose
Catalog # ROTH7168.2
Supplier:  Roth Carl
Marketsource
CAS Number:  50-69-1
D-(-)-Ribose

Glass bottle for solids, 100 g

CAS Number:  50-69-1
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:12,08371
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:90…95 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.