1,2-Dichloroethane
Glass bottle, 2,5 L
Catalog # ROTH6837.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Ethylene chloride, Ethylene dichloride, Dichloroethane
- CAS Number:107-06-2
- Formula:ClH₂CCH₂Cl
- MDL Number:MFCD00000963
- Molecular Weight:98.96 g/mol
- Merck Index:13,03831
- Boiling Pt:83.5 °C (1013 hPa)
- Melting Pt:–35.5 °C
- Density:1.254 g/cm³ (20 °C)
- Flash Pt:13 °C
- Storage Temperature:Ambient
- EINECS:203-458-1
- UN:1184
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



