Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Dextrin, white

Glass bottle for solids, 1 kg
Catalog # ROTH6778.1
Marketsource
Supplier:   Roth Carl
Dextrin, white
Catalog # ROTH6778.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9004-53-9
Dextrin, white

Glass bottle for solids, 1 kg

CAS Number:  9004-53-9
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-675-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications