Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 25%

Ammonia solution 25%

Glass bottle, 1 L
Catalog # ROTH6774.1
Corrosif
Irritant
Environnement
Marketsource
Supplier:   Roth Carl
Ammonia solution 25%
Catalog # ROTH6774.1
Supplier:  Roth Carl
Marketsource
CAS Number:  1336-21-6
Ammonia solution 25%

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
Demander un prix
Connexion Pour la disponibilité
1 * 1 L
Product Details & Documents
Synonymes:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • Numéro CAS:1336-21-6
  • Formule:NH₃
  • Numéro MDL:MFCD00066650
  • Poids moléculaire:35.05 g/mol
  • Point d'ébullition:32…37.7 °C (1013 hPa)
  • Point de fusion:–60…–57.5 °C
  • Densité:0.88…0.91 g/cm³ (20 °C)
  • Température de stockage:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III

Documents

Spécifications

Spécifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.