Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB)

3,3',5,5'-Tetramethylbenzidine (TMB)

Glass bottle for solids, 25 g
Catalog # ROTH6350.3
Marketsource
Supplier:   Roth Carl
3,3',5,5'-Tetramethylbenzidine (TMB)
Catalog # ROTH6350.3
Supplier:  Roth Carl
Marketsource
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB)

Glass bottle for solids, 25 g

CAS Number:  54827-17-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:166…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.