Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB)

Glass bottle for solids, 1 g
Catalog # ROTH6350.1
Marketsource
Supplier:   Roth Carl
3,3',5,5'-Tetramethylbenzidine (TMB)
Catalog # ROTH6350.1
Supplier:  Roth Carl
Marketsource
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB)

Glass bottle for solids, 1 g

CAS Number:  54827-17-7
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:166…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications