Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bacitracin

Bacitracin

Glass bottle for solids, 5 g
Catalogus # ROTH5655.1
Marketsource
Leverancier:   Roth Carl
Bacitracin
Catalogus # ROTH5655.1
Leverancier:  Roth Carl
Marketsource
CAS-nummer:  1405-87-4
Bacitracin

Glass bottle for solids, 5 g

CAS-nummer:  1405-87-4
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
  • CAS-nummer:1405-87-4
  • Formule:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL-nummer:MFCD00062640
  • Molecular Weight:1421.60 g/mol
Specificaties

Specificaties

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specificaties