Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 25%

Ammonia solution 25%

10 L
Catalog # ROTH5460.3
Corrosive
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Ammonia solution 25%
Catalog # ROTH5460.3
Supplier:  Roth Carl
Marketsource
CAS Number:  1336-21-6
Ammonia solution 25%

10 L

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III

Documents

Specifications

Specifications

  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.