Formaldehyde 35% in aqueous solution
10 L
Catalog # ROTH4980.4
Marketsource
Supplier: Roth Carl
Formaldehyde 35% in aqueous solution
Catalog # ROTH4980.4
Supplier: Roth Carl
Marketsource
CAS Number: 50-00-0
Jump to:
Product Details & Documents
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Specifications
Specifications
- Pk:10 L



