Formaldehyde 35% in aqueous solution
Glass bottle, 1 L
Catalog # ROTH4980.1
Marketsource
Supplier: Roth Carl
Formaldehyde 35% in aqueous solution
Catalog # ROTH4980.1
Supplier: Roth Carl
Marketsource
CAS Number: 50-00-0
Aller à :
Product Details & Documents
Synonymes: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- Numéro CAS:50-00-0
- Formule:CH₂O
- Numéro MDL:MFCD00003274
- Poids moléculaire:30.03 g/mol
- Température de stockage:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Spécifications
Spécifications
- Pack type:Glass bottle
- Pk:1 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



