Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formaldehyde 37% in aqueous solution

Formaldehyde 37% in aqueous solution

Glass bottle, 2,5 L
Catalog # ROTH4979.2
Toxic
Corrosive
Health
Marketsource
Supplier:   Roth Carl
Formaldehyde 37% in aqueous solution
Catalog # ROTH4979.2
Supplier:  Roth Carl
Marketsource
CAS Number:  50-00-0
Formaldehyde 37% in aqueous solution

Glass bottle, 2,5 L

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:2209
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.